missing translation for 'onlineSavingsMsg'
Learn More

Solvent Blue 104, TRC

Product Code. 30136314
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136314 10 g 10g
30135237 50 g 50g
30010794 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136314 Supplier Toronto Research Chemicals Supplier No. TRCS67674310G
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-75-6
Molecular Formula C32 H30 N2 O2
Molecular Weight (g/mol) 474.59
IUPAC Name 1,4-bis(2,4,6-trimethylanilino)anthracene-9,10-dione
SMILES Cc1cc(C)c(Nc2ccc(Nc3c(C)cc(C)cc3C)c4C(=O)c5ccccc5C(=O)c24)c(C)c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C32H30N2O2/c1-17-13-19(3)29(20(4)14-17)33-25-11-12-26(34-30-21(5)15-18(2)16-22(30)6)28-27(25)31(35)23-9-7-8-10-24(23)32(28)36/h7-16,33-34H,1-6H3
Formula Weight 474.2307
Chemical Name or Material Solvent Blue 104
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.