missing translation for 'onlineSavingsMsg'
Learn More

Solvent Blue 101 (Technical Grade), TRC

Product Code. 30410913
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30410913 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410913 Supplier Toronto Research Chemicals Supplier No. TRCS6767601G
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Solvent Blue 101
CAS 6737-68-4
Molecular Formula C28 H22 N2 O2
Quantity 1 g
Synonym 1,4-Bis[(2-methylphenyl)amino]-9,10-anthracenedione, 1,4-Bis(2-methylanilino)anthraquinone, C.I. 615670, C.I. Solvent Blue 101, Hytherm Blue E, Morplas Blue E
InChI Formula InChI=1S/C28H22N2O2/c1-17-9-3-7-13-21(17)29-23-15-16-24(30-22-14-8-4-10-18(22)2)26-25(23)27(31)19-11-5-6-12-20(19)28(26)32/h3-16,29-30H,1-2H3
SMILES Cc1ccccc1Nc2ccc(Nc3ccccc3C)c4C(=O)c5ccccc5C(=O)c24
IUPAC Name 1,4-bis(2-methylanilino)anthracene-9,10-dione
Molecular Weight (g/mol) 418.486
Formula Weight 418.168
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.