missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ Solutions for perfectION™ Combined ISE

Recommended reference solution for replacing electrolyte in ion selective electrodes.

Brand:  METTLER TOLEDO™ 51344751

Code : 59

Additional Details : CAS Number : 7757-79-1 Weight : 0.50000kg


 View more versions of this product

Product Code. 12641296

  • £139.00 / 300mL
In Stock
to see stock.
Add to basket

Description

Description

  • Keep electrolyte levels high by refilling regularly – when the probe is submerged, the electrolyte level should always be higher than the sample level.
  • Routinely refilling or replacing the electrolyte solution is necessary to maintain good electrode performance.
  • Reference table above to ensure proper electrolyte is selected for accurate measurement readings.
Specifications

Specifications

perfectION Electrolyte BIon Electrolyte B
perfectION ion electrolyte B, 5 x 60 mL
Combined ISE Solution for perfectION
perfectION Electrode: Chloride, Cyanide, Lead, or Silver/Sulfide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • perfectION Ion Electrolyte B

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P273-Avoid release to the environment.

Supplemental information
  • MIXTURE LIST-Contains : potassium nitrate, silver nitrate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers