missing translation for 'onlineSavingsMsg'
Learn More

Solution Kit of References GY, 7x100mL solutions EU Pharmacopoeia, Fisher Chemical™

Product Code. 12946233 Shop All Fisher Chemical Products
Click to view available options
Quantity:
7 x 100 mL
Unit Size:
700mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Chemical Name or Material Solution Kit of References GY
Packaging Bottle
Physical Form Liquid
Quantity 7 x 100 mL
Grade Eu. Ph.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.