missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solution Kit Of References BY, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
700 mL
Unit Size:
700mL
Specifications
Specifications
| Quantity | 700 mL |
Safety and Handling
Product Identifier
- Degree of Coloration Liquids BY1 - BY7. As specified in table 2.2.2.-3. of the European Pharmacopoeia. (OP 011)
- Danger
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H350-May cause cancer.
- H360-May damage fertility or the unborn child.
- H412-Harmful to aquatic life with long lasting effects.
- P202-Do not handle until all safety precautions have been read and understood.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction