missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solution fixing for isoelectric focusing in polyacrylamide gel EU Pharmacopoeia, Fisher Chemical™
Shop All Fisher Chemical Products

Specifications
Specifications
| Packaging | Bottle |
| Purity Grade Notes | EU Ph Ref: 1138700 |
| Quantity | 500 mL |
| Grade | Eu. Ph. |
Safety and Handling
Product Identifier
- Fixing solution for isoelectric focusing in polyacrylamide gel. European Pharmacopoeia Ref: 1138700 (RP 242)
- Danger
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501-Dispose of contents/container to .
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?