missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Solution dNTPs: PCR Nucleotide Mix
Free from DNase, RNase, and nicking enzyme activity
Brand: Cytiva 28406562
Code : GH
Additional Details : Weight : 0.50000kg
Description
- Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
- Buffer-free and ready-to-use solutions in multiple formats
- Functionally tested for long PCR and sequencing
Order Info
Custom formulation, testing, packaging, and labeling are available
Specifications
Liquid | |
2 mM | |
1 mL | |
Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling |
PCR Nucleotide Mix dNTP Mix (2mM each A,C,G,T) | |
>99% | |
Colorless |