missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Solution dNTPs: PCR Nucleotide Mix

Product Code. 10258184 Shop All Cytiva Products
Click to view available options
Concentration:
2 mM
10 mM
25 mM
Quantity:
500 μL
500 μm
1 mL
Unit Size:
1 set
500µL
This item is not returnable. View return policy

Product Code. 10258184

Brand: Cytiva 28406560

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Free from DNase, RNase, and nicking enzyme activity

  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer-free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing

Order Info

Custom formulation, testing, packaging, and labeling are available

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
Product Type PCR Nucleotide Mix dNTP Mix (25mM each A,C,G,T)
Concentration 25 mM
Purity >99%
Quantity 500 μm
Color Colorless
For Use With (Application) Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.