missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solute carrier family 22 member 18 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-57503
This item is not returnable.
View return policy
Description
Solute carrier family 22 member 18 Polyclonal specifically detects Solute carrier family 22 member 18 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Solute carrier family 22 member 18 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| BWR1AORCTL2, BWSCR1Asolute carrier family 22 (organic cation transporter), member 1-like, Efflux transporter-like protein, HET, imprinted multi-membrane spanning polyspecific transporter-related protein 1, Imprinted multi-membrane-spanning polyspecific transporter-related protein 1, IMPT1DKFZp667A184, ITMp45-BWR1A, ORCTL-2, organic cation transporter-like 2, Organic cation transporter-like protein 2, p45 Beckwith-Wiedemann region 1A, SLC22A1Lcandidate A, solute carrier family 22 member 18, Solute carrier family 22 member 1-like, solute carrier family 22, member 18, TSSC5p45-Beckwith-Wiedemann region 1 A, Tumor-suppressing STF cDNA 5 protein, Tumor-suppressing subchromosomal transferable fragment candidate gene 5 protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 5002 | |
| Human | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| SLC22A18 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PASTKGAKTDAQAPLPGGPRASVFDLKAIASLLRLPDVPRI | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction