missing translation for 'onlineSavingsMsg'
Learn More

Solute carrier family 22 member 18 Antibody, Novus Biologicals™

Product Code. 18360449 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18360449 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18360449 Supplier Novus Biologicals Supplier No. H00005002B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

Solute carrier family 22 member 18 Polyclonal antibody specifically detects Solute carrier family 22 member 18 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Solute carrier family 22 member 18
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_002546.2
Gene Alias BWR1AORCTL2, BWSCR1Asolute carrier family 22 (organic cation transporter), member 1-like, Efflux transporter-like protein, HET, imprinted multi-membrane spanning polyspecific transporter-related protein 1, Imprinted multi-membrane-spanning polyspecific transporter-related protein 1, IMPT1DKFZp667A184, ITMp45-BWR1A, ORCTL-2, organic cation transporter-like 2, Organic cation transporter-like protein 2, p45 Beckwith-Wiedemann region 1A, SLC22A1Lcandidate A, solute carrier family 22 member 18, Solute carrier family 22 member 1-like, solute carrier family 22, member 18, TSSC5p45-Beckwith-Wiedemann region 1 A, Tumor-suppressing STF cDNA 5 protein, Tumor-suppressing subchromosomal transferable fragment candidate gene 5 protein
Host Species Mouse
Immunogen SLC22A18 (NP_002546.2, 1 a.a. - 424 a.a.) full-length human protein. MQGARAPRDQGQSPGRMSALGRSSVILLTYVLAATELTCLFMQFSIVPYLSRKLGLDSIAFGYLQTTFGVLQLLGGPVFGRFADQRGARAALTLSFLAALALYLLLAAASSPALPGVYLLFASRLPGALMHTLPAAQMVITDLSAPEERPAALGRLGLCFGVGVILGSLLGGTLVSAYGIQCPAILAALATLLGAVLSFTCIPASTKGAKTDAQAPLPGGPRASVFDLKAIASLLRLPDVPRIFLVKVASNCPTGLFMVMFSIISMDFFQLEAAQAGYLMSFFGLLQMVTQGLVIGQLSSHFSEEVLLRASVLVFIVVGLAMAWMSSVFHFCLLVPGLVFSLCTLNVVTDSMLIKAVSTSDTGTMLGLCASVQPLLRTLGPTVGGLLYRSFGVPVFGHVQVAINTLVLLVLWRKPMPQRKDKVR
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5002
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.