missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solute Carrier Family 2 (Facilitated Glucose Transporter), Member 2 Rabbit Anti-Human, Polyclonal, Abnova&Trade;
Rabbit Polyclonal Antibody
Brand: Abnova PAB3157.400uL
This item is not returnable.
View return policy
Description
solute carrier family 2 (facilitated glucose transporter), member 2 Polyclonal antibody specifically detects solute carrier family 2 (facilitated glucose transporter), member 2 in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
| solute carrier family 2 (facilitated glucose transporter), member 2 | |
| Polyclonal | |
| Western Blot (1:1000) Flow cytometry (1:10-50) The optimal working dilution should be determined by the end user. |
|
| GLUT2 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human SLC2A2. | |
| RUO | |
| 6514 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| In PBS (0.09% sodium azide) | |
| Rabbit | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction