Learn More
STEMCELL Technologies Soluble ICAM-1/CD54, Mouse, Recombinant
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
- Mouse soluble ICAM-1 (∼95kDa) is a recombinant protein derived from the cell adhesion molecule
- This membrane protein is expressed on many cell types and is upregulated upon cell activation
- It binds to LFA-1 and Mac-1 on leukocytes
- Cell adhesion mediated by these molecules is critical for a wide range of cellular interactions in the immune system, including leukocyte adhesion to endothelial cells and interaction between T cells and antigen-presenting cells
- The adhesion of leukocytes to ICAM-1 requires functional activation of LFA-1 or Mac-1 on leukocytes
- Purified sICAM-1 adsorbed to a solid support binds activated cells expressing LFA-1 or Mac-1
- This binding can be blocked by the monoclonal anti-ICAM-1 antibody (in mouse with clone YN1/1; Catalog #01503), anti-LFA-1 or anti-Mac-1, indicating a direct interaction of these molecules in cell adhesion
- Soluble ICAM is useful for the studies of the activation of leukocyte adhesion in immune responses and inflammatory reactions
Specifications
Specifications
| Quantity | 100 μg |
| Cell Type | Lymphocytes; T Cells |