missing translation for 'onlineSavingsMsg'
Learn More

Solketal, 97%

Product Code. 10448120 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10448120

Brand: Thermo Scientific Acros 153651000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-79-8 | C6H12O3 | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-79-8
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00063238
InChI Key RNVYQYLELCKWAN-UHFFFAOYSA-N
Synonym solketal, 2,2-dimethyl-1,3-dioxolane-4-methanol, 2,2-dimethyl-1,3-dioxolan-4-yl methanol, glycerolacetone, dioxolan, isopropylidene glycerol, 1,2-isopropylideneglycerol, 1,2-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl, glycerol dimethylketal
PubChem CID 7528
IUPAC Name (2,2-dimethyl-1,3-dioxolan-4-yl)methanol
SMILES CC1(OCC(O1)CO)C

Specifications

Melting Point -50°C
Color Colorless
Density 1.0600g/mL
Boiling Point 188.0°C to 189.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4330 to 1.4350
Quantity 100 g
Beilstein 19, V,2, 362
Merck Index 15, 5259
Specific Gravity 1.06
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol, acetals, esters, ethers,, aromatic hydrocarbons, chlorinated hydrocarbons,, gasolines and petr.ether
Viscosity 11 mPa.s (20°C)
Formula Weight 132.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Solketal, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.