missing translation for 'onlineSavingsMsg'
Learn More

Solithromycin, MedChemExpress

Product Code. 16453437
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16453437

Brand: Medchem Express HY17593

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Solithromycin (CEM-101) is an orally bioavailable, effective antimicrobial agent, with IC50s for inhibition of cell viability, protein synthesis, and growth rate are 7.5 ng/mL, 40 ng/mL, and 125 ng/mL for Streptococcus pneumonia, Staphylococcus aureus, and Haemophilus influenzae, respectively. Solithromycin binds to the large 50S subunit of the ribosome and inhibits protein biosynthesis.

Solithromycin (CEM-101) is an orally bioavailable, effective antimicrobial agent, with IC50s for inhibition of cell viability, protein synthesis, and growth rate are 7.5 ng/mL, 40 ng/mL, and 125 ng/mL for Streptococcus pneumonia, Staphylococcus aureus, and Haemophilus influenzae, respectively. Solithromycin binds to the large 50S subunit of the ribosome and inhibits protein biosynthesis.

Specifications

Chemical Name or Material Solithromycin
Physical Form Solid
CAS 760981-83-7
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C43H65FN6O10
For Use With (Application) COVID-19-immunoregulation
Synonym CEM-101 OP-1068
Solubility Information DMSO : ≥ 32 mg/mL (37.87 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C[C@]([C@H](OC([C@@](F)(C([C@H](C)[C@H]1O[C@@](O[C@H](C)C[C@@H]2N(C)C)([H])[C@@H]2O)=O)C)=O)CC)(O3)[C@]([C@H](C([C@@H](C[C@@]1(C)OC)C)=O)C)([H])N(CCCCN4C=C(C5=CC(N)=CC=C5)N=N4)C3=O
Molecular Weight (g/mol) 845.01
Formula Weight 845.01
Percent Purity 98.87%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.