missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solithromycin, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Solithromycin (CEM-101) is an orally bioavailable, effective antimicrobial agent, with IC50s for inhibition of cell viability, protein synthesis, and growth rate are 7.5 ng/mL, 40 ng/mL, and 125 ng/mL for Streptococcus pneumonia, Staphylococcus aureus, and Haemophilus influenzae, respectively. Solithromycin binds to the large 50S subunit of the ribosome and inhibits protein biosynthesis.
Specifications
Specifications
| Chemical Name or Material | Solithromycin |
| Physical Form | Solid |
| CAS | 760981-83-7 |
| Color | White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C43H65FN6O10 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | CEM-101 OP-1068 |
| Solubility Information | DMSO : ≥ 32 mg/mL (37.87 mM) ∣H2O : < 0.1 mg/mL (insoluble) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction