missing translation for 'onlineSavingsMsg'
Learn More

Solifenacin Succinate, MedChemExpress

Product Code. 16494137
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16494137

Brand: Medchem Express HYA0002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Solifenacin Succinate (YM905) is a novel muscarinic receptor antagonist with pKis of 7.6, 6.9 and 8.0 for M1, M2 and M3 receptors, respectively.

Solifenacin Succinate (YM905) is a novel muscarinic receptor antagonist with pKis of 7.6, 6.9 and 8.0 for M1, M2 and M3 receptors, respectively.

Specifications

Chemical Name or Material Solifenacin Succinate
CAS 242478-38-2
Color White
Purity Grade Notes Research
Molecular Formula C27H32N2O6
Quantity 50 mg
Synonym YM905
Solubility Information H2O : 100 mg/mL (208.09 mM; Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (208.09 mM)
SMILES O=C(O[C@H]1CN2CCC1CC2)N3[C@H](C4=C(CC3)C=CC=C4)C5=CC=CC=C5.O=C(CCC(O)=O)O
Molecular Weight (g/mol) 480.55
Formula Weight 480.55
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.