missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solifenacin, MedChemExpress
Description
Solifenacin (YM905 free base) is a novel muscarinic receptor antagonist with pKis of 7.6, 6.9 and 8.0 for M1, M2 and M3 receptors, respectively.
Specifications
Specifications
| Chemical Name or Material | Solifenacin |
| CAS | 242478-37-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H26N2O2 |
| Quantity | 50 mg |
| Synonym | YM905 free base |
| Solubility Information | DMSO : ≥ 50 mg/mL (137.95 mM) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O=C(O[C@@]1([H])C[N@]2CC[C@@H]1CC2)N3CCC4=CC=CC=C4[C@@H]3C5=CC=CC=C5 |
| Molecular Weight (g/mol) | 362.46 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?