Learn More
Specac™ Solids Holder for Heating Jackets
Product Code. 15270238
Encompass_Preferred
Change view
Click to view available options
Diameter (Metric):
12 to 28 mm
Product Code. Diameter (Metric)
15270238 12 to 28 mm
1 options
Product Code. 15270238 Supplier Specac™ Supplier No. GS20600
Only null left
Add to basket
Add to basket
This Solids Holder is used with the Electrical Heating Jacket GS20730. The body has a fixed size aperture of 10mm and hence solid samples can be analysed from 12mm to 28mm diameter and up to 3mm thick. Does not require any windows.
Specifications
Diameter (Metric) 12 to 28 mm