Learn More
Fisherbrand™ Solid Rubber Stoppers
Offers higher resilience, flexibility and can withstand many chemical agents in normal laboratory use.
Brand:  Fisherbrand™ FB52015
Product Code. 10223371
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Temperature range -40° to +70°C
  • Not suitable for use with petroleum oils and hydrocarbon solvents