missing translation for 'onlineSavingsMsg'
Learn More

Solder, Lead-Free, ARMI∣MBH

Product Code. 17921446
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17921446 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17921446 Supplier ARMI/MBH Supplier No. MBH74X CA621G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.

Solder, Lead-Free, Ø40x15mm
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphorus/Aluminium/Selenium/Zinc/Cadmium/Mercury/Gold/Iron/Bismuth/Antimony/Arsenic/Nickel/Lead/Indium/Silver/Copper/Tin
CAS 7440-31-5
Quantity 1 Ea.
Form Disc
Concentration or Composition (by Analyte or Components) Phosphorus -21 ppm/Aluminium -8 ppm/Selenium -6 ppm/Zinc 0.0005 %/Cadmium 0.0007 %/Mercury 0.0042 %/Gold 0.0046 %/Iron 0.0098 %/Bismuth 0.011 %/Antimony 0.012 %/Arsenic 0.014 %/Nickel 0.0242 %/Lead 0.026 %/Indium 0.028 %/Silver 0.32 %/Copper 0.624 %
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.