missing translation for 'onlineSavingsMsg'
Learn More

Solabegron, MedChemExpress

Product Code. 16477768
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16477768 10 mg 10mg
16420238 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16477768 Supplier Medchem Express Supplier No. HY19436
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Solabegron (GW 427353) is a selective β3-adrenergic receptor agonist, stimulating cAMP accumulation in Chinese hamster ovary cells expressing the human β3-AR, with an EC50 value of 22 nM. Solabegron (GW 427353) is being developed for the treatment of overactive bladder and irritable bowel syndrome.

Solabegron (GW 427353) is a selective β3-adrenergic receptor agonist, stimulating cAMP accumulation in Chinese hamster ovary cells expressing the human β3-AR, with an EC50 value of 22 nM. Solabegron (GW 427353) is being developed for the treatment of overactive bladder and irritable bowel syndrome.

Specifications

Chemical Name or Material Solabegron
Physical Form Powder
CAS 252920-94-8
Color Light Brown
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C23H23ClN2O3
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym GW 427353
Solubility Information DMSO : ≥ 33.33 mg/mL (81.12 mM)
SMILES O=C(C1=CC(C2=CC=CC(NCCNC[C@@H](C3=CC=CC(Cl)=C3)O)=C2)=CC=C1)O
Molecular Weight (g/mol) 410.89
Formula Weight 410.89
Percent Purity 98.14%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.