Learn More
Thermo Scientific™ SOLA™ SPE Plates
Innovative Thermo Scientific™ SOLA™ Solid Phase Extraction Plates deliver clean, highly reproducible sample extracts with lower elution volumes—for greater sensitivity, reliability and cost savings.
Brand: Thermo Scientific™ 60309-001
262.93 GBP valid until 2024-10-31
Use promo code "21692" to get your promotional price.
Description
Obtain clean, highly reproducible sample extracts with lower elution volumes using Thermo Scientific™ SOLA™ Solid Phase Extraction (SPE) Plates. The enhanced performance gives you greater confidence in your analytical results and lowers operating costs—without compromising ease of use or requiring complex method development. Thanks to an innovative and unique frit-less design, SOLA SPE Plates deliver unparalleled performance, including:
- Higher levels of reproducibility
- Lower elution volumes
- High extract cleanliness
10mg/2mL 96 well plate format
Hydrophobic Reversed Phase (HRP), Strong Cation Exchanger (SCX) and Strong Anion Exchanger (SAX) phase types
SOLA products are also available in cartridge format.
Specifications
10 mg/2 mL; SOLA 96-well SPE plate; 1 Pk. | |
Bioanalytical Applications - Hydrophobic Retention of Neutral Compounds with Complementary Retention of Polar Analytes. An All-Purpose Phase. | |
HRP (Reversed Phase Polymeric) | |
2 mL |
10 mg | |
SOLA | |
96-well SPE Plate |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.