missing translation for 'onlineSavingsMsg'
Learn More

Sol8 (mouse myoblast) Whole cell lysate, Non-denatured, Abnova

Non-denatured whole cell lysate of Sol8 (mouse myoblast)

Brand:  Abnova L058V2.200ug

Additional Details : Weight : 0.02000kg

Product Code. 16079635

  • £91.00 / 200µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Tissue: Skeletal Muscle
  • Host: Mouse
  • Lysis Buffer: Modified RIPA Lysis Buffer: 50mM Tris-HCl pH 7.4, 150mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF
  • Storage buffer: In modified RIPA Lysis Buffer

Western Blotting, Immunoprecipitation

Specifications

Specifications

Immunoprecipitation, Western Blot
Whole cell lysate (non-denatured)
Cell lysate was prepared by homogenization in ice-cold modified RIPA Lysis Buffer with cocktail of protease inhibitors (Sigma). Cell debris was removed by centrifugation. Protein concentration was determined with Bradford assay (Bio-Rad protein assay, Microplate Standard assay).
200 μg
Mouse
Skeletal Muscle
Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF.
12.5% SDS-PAGE Stained with Coomassie Blue.
In modified RIPA Lysis Buffer.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers