missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOHLH1 Polyclonal Antibody
SOHLH1 Polyclonal Antibody
Brand: Invitrogen PA566258
This item is not returnable.
View return policy
Description
SOHLH1 Polyclonal Antibody for ICC/IF
Specifications
| SOHLH1 | |
| Polyclonal | |
| Unconjugated | |
| SOHLH1 | |
| bA100C15.3; bHLHe80; C9orf157; Gm110; helix-loop-helix protein NOHLH; newborn ovary helix loop helix; NOHLH; RGD1564440; RP11-100C15.3; Sohlh1; SPATA27; spermatogenesis and oogenesis specific basic helix-loop-helix 1; spermatogenesis- and oogenesis-specific basic helix-loop-helix-containing protein 1; spermatogenesis associated 27; TEB2; Tohlh1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 402381 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5JUK2 | |
| SOHLH1 | |
| Recombinant Human SOHLH1. Recombinant protein control fragment (Product #RP-106939). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction