missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOGA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546498
This item is not returnable.
View return policy
Description
Peptide sequence: EPGDPEKDTK EKPGLSSRDC NHLGALACQD PPGRQMQRSY TAPDKTGIRV Sequence homology: Guinea Pig: 82%; Human: 100%; Rat: 77%.
SOGA1 regulates autophagy by playing a role in the reduction of glucose production in an adiponectin- and insulin-dependent manner.
Specifications
| SOGA1 | |
| Polyclonal | |
| Unconjugated | |
| Soga1 | |
| 80-kDa SOGA fragment; 9830001H06Rik; AI426038; C20orf117; C-terminal 80 kDa form; D430036N24Rik; dJ132F21.1; KIAA0889; N-terminal form; Protein SOGA1; Soga; SOGA family member 1; Soga1; Suppressor of glucose by autophagy; suppressor of glucose from autophagy; suppressor of glucose, autophagy associated 1; suppressor of glucose, autophagy-associated protein 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 140710 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O94964 | |
| Soga1 | |
| Synthetic peptide directed towards the C-terminal of human SOGA1 (aa 965-1014). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction