Learn More
Syngene™ Dymension 2 Software
GREENER_CHOICE
Product Code. 12847004 Shop All Syngene Products
Change view
Click to view available options
For Use With (Equipment):
Gbox-Chemi-XX6/9
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
12847004 Gbox-Chemi-XX6/9 Each
1 options
Product Code. 12847004 Supplier Syngene™ Supplier No. DYM2
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
Syngene™ Dymension 2 Software is a 2D analysis software for gel electrophoresis.
  • Software for the rapid analysis of 2D protein spots from multiple gels
  • With interactive process flow diagram
  • Main pane and secondary panes for viewing replicates and samples
  • Synchronised zoom with intensity autoscale
  • Automatic and precise image warping with vector view
  • Manual warp editing
  • Automatic background subtraction and noise filtering
  • SYPRO Ruby crystal filter
  • Automatic spot detection Region of interest option
  • Gaussian spot modelling option
  • 3 x 10MB images spot detected in eight seconds
  • Spot detection with user adjustable parameters
  • Spot editing, filtering and normalization
  • Spot matching across multiple gels
  • Manual match editing pI and MW ladder calibration
  • Main pane and secondary panes for replicates and samples
  • Bar chart 3D spot profile Matched spot plot
  • Correlation plot Table of spot matching results
  • Scatter plot Statistical tests
  • Export to Word and Excel
  • Spot Picker interface
  • Supplied with one years' support
Specifications
For Use With (Application) 2d electrophoresis analysis
For Use With (Equipment) Gbox-Chemi-XX6/9