Learn More
Memmert™ Software Conforming To Fda Celsius Fda Edition, Base License For 1 Unit, For Ttc256
Shop All Memmert Products
Click to view available options
Includes:
Base License for 1 Unit
Unit Size:
Each
Description
- For use with Memmert ovens
Specifications
Specifications
| Certifications/Compliance | 21 CFR Part 11, FDA |
| For Use With (Equipment) | Use of Electronically Stored Data Sets and Electronic Signatures |
| Type | Software |
| Includes | Base License for 1 Unit |