missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Software AtmoCONTROL for UN110plus

Product Code. 17559825 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN110plus Oven
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17559825

Brand: Memmert™ UN110plusFDAQ1

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

AtmoCONTROL Software meets the requirements for the use of electronically stored data sets and electronic signatures.

  • Software conforming to FDA AtmoCONTROL
  • Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
  • Base licence for the control of one unit
  • Respective IQ/OQ documents available in German and English language (without surcharge)

Specifications

Certifications/Compliance Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
For Use With (Equipment) UN110plus Oven
Type Software
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.