Learn More
Hamilton™ SoftGrip™ Fixed Volume Pipette
Click to view available options
Volume (Metric):
5 μL
10 μL
50 μL
100 μL
200 μL
250 μL
300 μL
500 μL
1000 μL
Color:
Brick Red
Burnt Orange
Mustard
Olive Green
Purple
Sandstone
Sky Blue
Steel Blue
Violet
Unit Size:
Each
Description
- Wide range of volumes available
- Ejection sleeve made from a high strength, chemically resistant polymer
- Universal calibration key that reduces the need for costly calibration services
- Calibration takes only minutes to perform and eliminates downtime
- Tip nozzle ensures a proper fit with universal pipette tips
- Unique color coding and labeling clearly indicates the volume of the pipettes
Specifications
Specifications
| Accuracy | 0.40±% |
| Autoclavable | Autoclavable |
| Color | Burnt Orange |
| Number of Channels | 1 |
| Type | Fixed Volume Pipette |
| Volume (Metric) | 250 μL |
| Precision | 0.18±% |
| Quantity | 1/Pk. |