missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ SoftFit-L™ Empty Racks for use with SoftFit-L Reload Inserts and Reload System Towers

Click to view available options
For Use With (Equipment):
20 μL Pipet Tip, 200 μL Pipet Tip, 300 μL Pipet Tip
Rainin LTS LiteTouch
Quantity:
10 Racks/Pk., 5 Pks./Cs.
8 Racks/Pk., 4 Pks./Cs.
Unit Size:
Pack of 10
Pack of 8
Description
Thermo Scientific™ART SoftFit-L™ empty racks, when used in conjunction with SoftFit-L reload towers and inserts, offer an environmentally friendly packaging option with less plastic waste than traditional racks. Compatible with ART SoftFit-L reload inserts and reload towers.
Specifications
Specifications
| Volume (Metric) | 1000 μL |
| Model | SoftFit L Rack |
| Clean Claims | DNase, RNase, DNA, ATP, and Endotoxin Free |
| Format | Hinged Rack |
| Quantity | 8 Racks/Pk., 4 Pks./Cs. |
| For Use With (Equipment) | Rainin LTS LiteTouch |
| Product Line | ART SoftFit |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering