missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ SoftGrip™ Adjustable Volume Single Channel Pipette

Product Code. 10395112
Click to view available options
Volume (Metric):
0.2 to 2 μL
1 to 10 μL
10 to 100 μL
100 to 1000 μL
2.5 to 25 μL
30 to 300 μL
Color:
Aqua
Blue
Brick Red
Green
Purple
Violet
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10395112

Brand: Hamilton™ 5501940

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

With improved volume adjustment accuracy by removing parallax errors.

  • Available in a wide range of volumes
  • Isolated volume adjustment
  • Dual indicator
  • Slim ejection sleeve manufactured from high-strength, chemically resistant polymer
  • Slim design allows the pipette to reach the bottom of narrow tubes without extended tips
  • Universal calibration key that reduces the need for costly calibration services
  • Calibration requires only minutes to perform
  • Tip nozzle ensures a proper fit with universal pipette tips
  • Unique color coding and labeling clearly indicates the working range of the pipettes

Specifications

Accuracy 1.60 ±% at 10°C
Autoclavable Autoclavable
Color Blue
For Use With Universal Pipette Tip
Grip Type Soft
Number of Channels 1
Product Line SoftGrip
Type Soft Grip Microliter Pipette
Volume (Metric) 100 to 1000 μL
Increments 2.0μL
Precision 0.12 ±% at 100%
Quantity 1 Pack
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.