missing translation for 'onlineSavingsMsg'
Learn More

Sofosbuvir impurity L, MedChemExpress

Product Code. p-200097474
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16431258

Brand: Medchem Express HYI1196

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sofosbuvir impurity L, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.

Sofosbuvir impurity L, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.

Specifications

Chemical Name or Material Sofosbuvir impurity L
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H29FN3O10P
Quantity 5 mg
SMILES O=[P@@](N[C@@H](C)C(OC(C)C)=O)(OC[C@@H]1[C@@H](O)[C@](F)(C)[C@H](N2C=CC(N(O)C2=O)=O)O1)OC3=CC=CC=C3
Molecular Weight (g/mol) 545.45
Formula Weight 545.45
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.