missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sofosbuvir impurity I, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
Sofosbuvir impurity I, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.
Specifications
Specifications
| Chemical Name or Material | Sofosbuvir impurity I |
| CAS | 2164516-85-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H27FN3O9P |
| Quantity | 5 mg |
| SMILES | O=[P@@](N[C@@H](C)C(OCC)=O)(OC[C@@H]1[C@@H](O)[C@](F)(C)[C@H](N2C=CC(NC2=O)=O)O1)OC3=CC=CC=C3 |
| Molecular Weight (g/mol) | 515.43 |
| Formula Weight | 515.43 |
| Grade | Research |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction