missing translation for 'onlineSavingsMsg'
Learn More

Sofosbuvir impurity A, MedChemExpress

Product Code. 16438877
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438877 5 mg 5mg
16431469 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16438877 Supplier Medchem Express Supplier No. HY15005C
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sofosbuvir impurity A, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.

Sofosbuvir impurity A, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.

Specifications

Chemical Name or Material Sofosbuvir impurity A
CAS 1496552-16-9
Color White
Purity Grade Notes Research
Molecular Formula C22H29FN3O9P
Quantity 5 mg
SMILES O=[P@](N[C@H](C)C(OC(C)C)=O)(OC[C@@H]1[C@@H](O)[C@](F)(C)[C@H](N2C=CC(NC2=O)=O)O1)OC3=CC=CC=C3
Molecular Weight (g/mol) 529.45
Formula Weight 529.45
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.