missing translation for 'onlineSavingsMsg'
Learn More

Sofiniclin, MedChemExpress

Product Code. 16478787
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16478787

Brand: Medchem Express HY14824

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sofiniclin (ABT 894), an agonist of nicotinic acetylcholine receptor (nAChR), is used as a potential non-stimulant research for attention-deficit/hyperactivity disorder (ADHD).

Sofiniclin (ABT 894), an agonist of nicotinic acetylcholine receptor (nAChR), is used as a potential non-stimulant research for attention-deficit/hyperactivity disorder (ADHD).

Specifications

Chemical Name or Material Sofiniclin
CAS 799279-80-4
Color Light Nude
Purity Grade Notes Research
Molecular Formula C10H11Cl2N3
Quantity 5 mg
Synonym ABT 894
Solubility Information DMSO : 35.71 mg/mL (146.28 mM; ultrasonic and warming and heat to 60°C)
SMILES ClC1=C(Cl)C=C(N2C[C@]3([H])CN[C@]3([H])C2)C=N1
Molecular Weight (g/mol) 244.12
Formula Weight 244.12
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.