missing translation for 'onlineSavingsMsg'
Learn More

Sofalcone, MedChemExpress

Product Code. 16491148
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491148 5 g 5g
16482379 1 g 1g
16482306 500 mg 500mg
16458866 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491148 Supplier Medchem Express Supplier No. HYB2184
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sofalcone, a gastric antiulcer agent, is known to induce the expression of Heme oxygenase-1 (HO-1) in gastric epithelium.

Sofalcone, a gastric antiulcer agent, is known to induce the expression of Heme oxygenase-1 (HO-1) in gastric epithelium.

Specifications

Chemical Name or Material Sofalcone
CAS 64506-49-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H30O6
Quantity 5 g
Solubility Information DMSO : ≥ 100 mg/mL (221.97 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(O)COC1=CC(OC/C=C(C)/C)=CC=C1C(/C=C/C2=CC=C(OC/C=C(C)/C)C=C2)=O
Molecular Weight (g/mol) 450.52
Formula Weight 450.52
Percent Purity 99.12%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.