missing translation for 'onlineSavingsMsg'
Learn More

Sodium Valproate, TRC

Product Code. 30135032
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135032 25 g 25g
30138668 10 g 10g
30041368 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135032 Supplier Toronto Research Chemicals Supplier No. TRCS67450025G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1069-66-5
Molecular Formula C8 H15 O2 . Na
Molecular Weight (g/mol) 166.19
Synonym Sodium Valproate, 2-Propylpentanoic acid sodium salt (1:1), 2-Propylvaleric acid sodium salt, Acediprol, Convulex, Depacon, Depakene, Depakene R, Depakene syrup 8, Depakin, Depakine chronosphere, Dipropylacetate sodium, Epilim, KW 6066N, Orfiril, Selenica R, Sodium α,α-dipropylacetate, Sodium 2-propylpentanoate, Sodium 2-propylvalerate, Sodium bispropylacetate, Sodium dipropylacetate, Sodium valproate, Valerin, Valproate sodium
IUPAC Name sodium;2-propylpentanoate
SMILES [Na+].CCCC(CCC)C(=O)[O-]

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H16O2.Na/c1-3-5-7(6-4-2)8(9)10;/h7H,3-6H2,1-2H3,(H,9,10);/q;+1/p-1
Formula Weight 166.097
Percent Purity >95%
Chemical Name or Material Sodium Valproate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.