missing translation for 'onlineSavingsMsg'
Learn More

Sodium Valproate, LoGiCal

Product Code. 30461803
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461803 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461803 Supplier LoGiCal Supplier No. LGCFOR0751.11
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Sodium Valproate
CAS 1069-66-5
Analyte or Component Names Sodium Valproate
Shipping Condition Room Temperature
Molecular Formula C8 H15 O2 . Na
Certifications/Compliance ISO 9001
Synonym Sodium Valproate, 2-Propylpentanoic acid sodium salt (1:1), 2-Propylvaleric acid sodium salt, Acediprol, Convulex, Depacon, Depakene, Depakene R, Depakene syrup 8, Depakin, Depakine chronosphere, Dipropylacetate sodium, Epilim, KW 6066N, Orfiril, Selenica R, Sodium a,a-dipropylacetate, Sodium 2-propylpentanoate, Sodium 2-propylvalerate, Sodium bispropylacetate, Sodium dipropylacetate, Sodium valproate, Valerin, Valproate sodium, Sodium a,a-dipropylacetate
InChI Formula InChI=1S/C8H16O2.Na/c1-3-5-7(6-4-2)8(9)10;/h7H,3-6H2,1-2H3,(H,9,10);/q;+1/p-1
Solution Type Neat
SMILES [Na+].CCCC(CCC)C(=O)[O-]
IUPAC Name sodium;2-propylpentanoate
Industry Type Clinical Testing
Molecular Weight (g/mol) 166.19
Quantity 10 mg
Formula Weight 166.097 g/mol
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.