missing translation for 'onlineSavingsMsg'
Learn More

Sodium valproate, 99.9%, MP Biomedicals™

Product Code. 11474361 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474361 10 g 10g
11484361 25 g 25g
11494361 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474361 Supplier MP Biomedicals Supplier No. 0215206410
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1069-66-5
Molecular Formula C8H15NaO2
Molecular Weight (g/mol) 166.196
InChI Key AEQFSUDEHCCHBT-UHFFFAOYSA-M
Synonym sodium valproate, sodium 2-propylpentanoate, valproate sodium, valproic acid sodium salt, eurekene, labazene, orfiril, epilim, valproic acid sodium, valerin
PubChem CID 16760703
ChEBI CHEBI:9925
IUPAC Name sodium;2-propylpentanoate
SMILES CCCC(CCC)C(=O)[O-].[Na+]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 10 g
Assay Percent Range 0.999
Formula Weight 166.2
Chemical Name or Material Sodium valproate
compliance-icons
Product Identifier
  • Sodium Valproate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sodium Valproate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.