missing translation for 'onlineSavingsMsg'
Learn More

Sodium tungsten oxide dihydrate, ACS, 99.0-101.0%, Thermo Scientific Chemicals

Catalog Number 11397948
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10213-10-2
H4Na2O6W
329.846
MFCD00149190
WPZFLQRLSGVIAA-UHFFFAOYSA-N
sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o
150191
CHEBI:63939
disodium;dioxido(dioxo)tungsten;dihydrate
O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10213-10-2
H4Na2O6W
329.846
MFCD00149190
WPZFLQRLSGVIAA-UHFFFAOYSA-N
sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o
150191
CHEBI:63939
disodium;dioxido(dioxo)tungsten;dihydrate
O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+]

In organic chemistry, sodium tungstate is used as catalyst for epoxidation of alkenes and oxidation of alcohols into aldehydes or ketones. Used in examining the serum protein, serum protein-free filtrate. Used as alkaloid precipitating agent. Used in manufacture of metal tungsten, tungstate and tungsten oxide. Waterproof and fireproof fabric.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 100°C (in -2H2O)
Color White
Physical Form Powder
Quantity 100 g
Merck Index 14,8698
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 329.86 (293.83 Anhydrous)
Percent Purity 99.0 to 101.0%
Grade ACS
Odor Odorless
Density 3.250 g/mL
Chemical Name or Material Sodium tungsten oxide dihydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'