missing translation for 'onlineSavingsMsg'
Learn More

Sodium tripolyphosphate, Thermo Scientific Chemicals

Catalog Number 11366778
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7758-29-4
Na5O10P3
367.86
MFCD00003514
HWGNBUXHKFFFIH-UHFFFAOYSA-I
sodium tripolyphosphate, sodium triphosphate, pentasodium triphosphate, stpp, thermphos, armofos, polygon, tripoly, rhodiaphos lv, thermphos n
24455
pentasodium;[oxido(phosphonatooxy)phosphoryl] phosphate
[O-]P(=O)([O-])OP(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+].[Na+]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7758-29-4
Na5O10P3
367.86
MFCD00003514
HWGNBUXHKFFFIH-UHFFFAOYSA-I
sodium tripolyphosphate, sodium triphosphate, pentasodium triphosphate, stpp, thermphos, armofos, polygon, tripoly, rhodiaphos lv, thermphos n
24455
pentasodium;[oxido(phosphonatooxy)phosphoryl] phosphate
[O-]P(=O)([O-])OP(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+].[Na+]

Sodium tripolyphosphate is used in water softening, heavy-duty fabric washing, automatic dish washing, drilling fluid cleaning and liquid cattle feed supplements. It is also used as a peptizing, emulsifying, dispersing agent and as a hydrogen peroxide solution stabilizer. It acts as a preservative for seafood, meats, poultry, and animal feeds. It finds application as a polyanion crosslinker in polysaccharide based drug delivery. Further, it is used as a sealant for leaking farm ponds and a calcium and magnesium chelating agent. In addition to this, it is used in ceramics, leather tanning, flame retardants, paper pulping, paper coatings, textile processing, ore flotation, anticorrosion pigments and rubber production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 622°C
Color White
Physical Form Granules
Quantity 500 g
Merck Index 14,8697
Solubility Information Soluble in water; Slightly soluble in methanol; Insoluble in diethyl ether and n-octanol.
Formula Weight 367.91
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Sodium tripolyphosphate
compliance-icons
Product Identifier
  • Sodium tripolyphosphate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Pentasodium triphosphate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'