missing translation for 'onlineSavingsMsg'
Learn More

Sodium trimethylsilanolate, 1M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10066338 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10066338 100 mL AcroSeal™ Glass Bottle 100mL
10509778 800 mL AcroSeal™ Glass Bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10066338 Supplier Thermo Scientific Acros Supplier No. 428841000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18027-10-6 , 75-09-2
Molecular Formula C3H9NaOSi
Molecular Weight (g/mol) 112.18
MDL Number MFCD00013926
InChI Key HSNUIYJWTSJUMS-UHFFFAOYSA-N
Synonym sodium trimethylsilanolate, silanol, trimethyl-, sodium salt, trimethylsilanol sodium salt, silanol, 1,1,1-trimethyl-, sodium salt 1:1, silanol,1,1,1-trimethyl-, sodium salt 1:1, naotms, sodiooxytrimethylsilane, naosi me 3, trimethylsilyloxy sodium, sodium trimethylsilanolat
PubChem CID 23672319
IUPAC Name sodium;trimethyl(oxido)silane
SMILES C[Si](C)(C)[O-].[Na+]

Specifications

Quantity 100 mL
Flash Point 63°C
Formula Weight 112.18
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.28
Density 1.2800g/mL
Chemical Name or Material Sodium trimethylsilanolate
compliance-icons compliance-icons
Product Identifier
  • Sodium trimethylsilanolate, 1M solution in dichloromethane
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H336-May cause drowsiness or dizziness.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.