missing translation for 'onlineSavingsMsg'
Learn More

Sodium trimetaphosphate

Product Code. 11319379
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319379 1 kg 1kg
11329379 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319379 Supplier Thermo Scientific Alfa Aesar Supplier No. 089063.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7785-84-4 | Na3O9P3 | 305.882 g/mol

Sodium trimetaphosphate is used as a crosslinking agent in the cross-linked amylase-resistant starch. It acts as an intermediate in food industries. Further, it serves as a corrosion inhibitors, anti-scaling agents, fillers, finishing agents, plating agents and surface treating agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7785-84-4
Molecular Formula Na3O9P3
Molecular Weight (g/mol) 305.882
MDL Number MFCD00867826
InChI Key UGTZMIPZNRIWHX-UHFFFAOYSA-K
Synonym sodium trimetaphosphate, trisodium trimetaphosphate, trisodium metaphosphate, cyclic sodium trimetaphosphate, unii-3ih6169rl0, sodium phosphate napo3 3, sodium trimetaphosphate usan, metaphosphoric acid, trisodium salt, sodium metaphosphate na3 p3o9, trisodium trimetaphosphate na3p3o9
PubChem CID 24579
IUPAC Name trisodium;2,4,6-trioxido-1,3,5,2$l^{5},4$l^{5},6$l^{5}-trioxatriphosphinane 2,4,6-trioxide
SMILES [O-]P1(=O)OP(=O)(OP(=O)(O1)[O-])[O-].[Na+].[Na+].[Na+]

Specifications

Physical Form Powder
Quantity 1 kg
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 305.89
Density 2.49
Chemical Name or Material Sodium trimetaphosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.