missing translation for 'onlineSavingsMsg'
Learn More

Sodium trifluoromethanesulfonate

Product Code. 11397967
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397967 10 g 10g
11307977 2 g 2g
11387967 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397967 Supplier Thermo Scientific Alfa Aesar Supplier No. 020837.09
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2926-30-9 | CF3NaO3S | 172.053 g/mol

Catalyzes the aza Diels-Alder reaction of Danishefsky's diene with imines in water under neutral conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2926-30-9
Molecular Formula CF3NaO3S
Molecular Weight (g/mol) 172.053
MDL Number MFCD00061607
InChI Key XGPOMXSYOKFBHS-UHFFFAOYSA-M
Synonym sodium trifluoromethanesulfonate, sodium triflate, trifluoromethanesulfonic acid sodium salt, sodium trifluoro methane sulfonate, methanesulfonic acid, trifluoro-, sodium salt, pubchem23598, cf3so3na, acmc-1cfau, sodiumtrifluoromethanesulfonate, ksc491a1d
PubChem CID 23676748
IUPAC Name sodium;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point 253°C to 255°C
Color White
Odor Odorless
Linear Formula CF3SO3Na
Quantity 10 g
Beilstein 3728797
Sensitivity Hygroscopic
Solubility Information Soluble in water (1000g/L).
Formula Weight 172.06
Physical Form Powder
Chemical Name or Material Sodium trifluoromethanesulfonate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium trifluoromethanesulfonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Sodium trifluoromethanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.