missing translation for 'onlineSavingsMsg'
Learn More

Sodium trifluoromethanesulfonate, 98%

Product Code. p-7025345
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435666 5 g 5g
11445666 25 g 25g
11455666 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435666 Supplier Thermo Scientific Alfa Aesar Supplier No. B20776.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2926-30-9 | CF3NaO3S | 172.053 g/mol

Sodium trifluoromethanesulfonate is used in the preparation of N-fluoro-2-methylpyridinium triflate by reaction with dinitrogen difluoride as a reagent. It is also used as a chaotropic mobile phase additive in reversed-phase liquid chromatography (RP-LC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium trifluoromethanesulfonate is used in the preparation of N-fluoro-2-methylpyridinium triflate by reaction with dinitrogen difluoride as a reagent. It is also used as a chaotropic mobile phase additive in reversed-phase liquid chromatography (RP-LC).

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2926-30-9
Molecular Formula CF3NaO3S
Molecular Weight (g/mol) 172.053
MDL Number MFCD00061607
InChI Key XGPOMXSYOKFBHS-UHFFFAOYSA-M
Synonym sodium trifluoromethanesulfonate, sodium triflate, trifluoromethanesulfonic acid sodium salt, sodium trifluoro methane sulfonate, methanesulfonic acid, trifluoro-, sodium salt, pubchem23598, cf3so3na, acmc-1cfau, sodiumtrifluoromethanesulfonate, ksc491a1d
PubChem CID 23676748
IUPAC Name sodium;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point 251°C to 255°C
Quantity 5 g
Beilstein 3728797
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 172.06
Percent Purity 98%
Chemical Name or Material Sodium trifluoromethanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.