missing translation for 'onlineSavingsMsg'
Learn More

Sodium Trifluoroacetate, TRC

Product Code. 30007393
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007393 2.5 g 2.5g
18070210 250 mg 250mg
17979849 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007393 Supplier Toronto Research Chemicals Supplier No. TRCS6550202.5G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2923-18-4
Molecular Formula C2 F3 O2 . Na
Molecular Weight (g/mol) 136.01
Synonym Acetic acid, 2,2,2-trifluoro-, sodium salt (1:1), Acetic acid, trifluoro-, sodium salt (8CI,9CI), Sodium 2,2,2-trifluoroacetate, Sodium perfluoroacetate, Sodium trifluoroacetate, Sodium trifluoromethanecarboxylate, Trifluoroacetate sodium, Trifluoroacetic acid, sodium salt, Trifluoroacetic Acid Sodium, Trifluoroacetic acid sodium, Sodium Trifluoroacetate, Sodium Trifluoroacetate
IUPAC Name sodium;2,2,2-trifluoroacetate
SMILES [Na+].[O-]C(=O)C(F)(F)F

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C2HF3O2.Na/c3-2(4,5)1(6)7;/h(H,6,7);/q;+1/p-1
Formula Weight 135.9748
Chemical Name or Material Sodium Trifluoroacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.