missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium trifluoroacetate, 98%, Thermo Scientific Chemicals
16.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Sodium trifluoroacetate is used in acid catalyzed reactions. It is also used as an intermediate for active pharmaceutical ingredients (API). Further, it acts as a precursor for the trifluoromethylation of aldehydes using copper(I) halide as a catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium trifluoroacetate is used in acid catalyzed reactions. It is also used as an intermediate for active pharmaceutical ingredients (API). Further, it acts as a precursor for the trifluoromethylation of aldehydes using copper(I) halide as a catalyst.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases. Hygroscopic. Moisture sensitive.
Sodium trifluoroacetate is used in acid catalyzed reactions. It is also used as an intermediate for active pharmaceutical ingredients (API). Further, it acts as a precursor for the trifluoromethylation of aldehydes using copper(I) halide as a catalyst.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases. Hygroscopic. Moisture sensitive.
Specifications
Melting Point | ∽207°C (decomposition) |
Assay Percent Range | 98% |
Quantity | 25 g |
Beilstein | 3597949 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
IUPAC Name | sodium;2,2,2-trifluoroacetate |
Formula Weight | 136 |
Percent Purity | 98% |
Chemical Name or Material | Sodium Trifluoroacetate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'