missing translation for 'onlineSavingsMsg'
Learn More

Sodium trichloroacetate, 97%

Product Code. 11403970
Change view
Click to view available options
Quantity:
10 g
250 g
1000 g
5000 g
Unit Size:
1000g
10g
250g
5000g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11403970 10 g 10g
11408856 250 g 250g
11498846 1000 g 1000g
11418856 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11403970 Supplier Thermo Scientific Alfa Aesar Supplier No. A17004.09
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 650-51-1 | C2Cl3NaO2 | 185.36 g/mol

Sodium trichloroacetate, is used in acid binding reagent in the textile industry in the dying process. Sodium trichloroacetate has been reported previously to be an effective denaturation reagent for proteins was applied to poly(l-lysine) and poly(l-glutamic acid) to see its effects on a coil-to-helix transition and on the chemical reactivities of the ε-amino group of poly(l-lysine). Addition of sodium trichloroacetate to poly-(l-lysine) induced a helical conformation even at neutral pH where the ε-amino group of the polymer was protonated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium trichloroacetate, is used in acid binding reagent in the textile industry in the dying process. Sodium trichloroacetate has been reported previously to be an effective denaturation reagent for proteins was applied to poly(l-lysine) and poly(l-glutamic acid) to see its effects on a coil-to-helix transition and on the chemical reactivities of the ε-amino group of poly(l-lysine). Addition of sodium trichloroacetate to poly-(l-lysine) induced a helical conformation even at neutral pH where the ε-amino group of the polymer was protonated.

Solubility
Soluble in water, ethanol, methanol, acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 650-51-1
Molecular Formula C2Cl3NaO2
Molecular Weight (g/mol) 185.36
MDL Number MFCD00064198
InChI Key SAQSTQBVENFSKT-UHFFFAOYSA-M
Synonym sodium trichloroacetate, tca-sodium, trichloroacetic acid sodium salt, antiperz, antyperz, tca sodium, acp grass killer, sodium tca, varitox, stca
PubChem CID 23681045
ChEBI CHEBI:81976
IUPAC Name sodium;2,2,2-trichloroacetate
SMILES C(=O)(C(Cl)(Cl)Cl)[O-].[Na+]

Specifications

Melting Point >300°C
Quantity 10 g
UN Number UN3077
Beilstein 3572664
Merck Index 14,9627
Formula Weight 185.37
Percent Purity 97%
Chemical Name or Material Sodium trichloroacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.