missing translation for 'onlineSavingsMsg'
Learn More

Sodium triacetoxyborohydride, 95%

Product Code. 11496400
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11496400

Brand: Thermo Scientific Alfa Aesar B22060.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56553-60-7 | C6H10BNaO6 | 211.94 g/mol

Sodium triacetoxyborohydride is used as a reagent in reductive amination of ketones, aldehydes and lactamization of carbonyl compounds with amines. It also serves as a reducing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium triacetoxyborohydride is used as a reagent in reductive amination of ketones, aldehydes and lactamization of carbonyl compounds with amines. It also serves as a reducing agent in organic synthesis.

Solubility
Soluble in dimethyl sulfoxide, methanol, benzene, toluene, terahydrofuran, dioxane and methylene chloride.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, water, alcohols and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56553-60-7
Molecular Formula C6H10BNaO6
Molecular Weight (g/mol) 211.94
MDL Number MFCD00012211
InChI Key HHYFEYBWNZJVFQ-UHFFFAOYSA-N
Synonym c6h10bnao6, sodium triacetoxyboron 1-, pubchem12871, sodium triacetoxy borohyride, sodium triacetyloxyboron 1-, tris acetoxy hydroborate sodium salt, sodium bis acetyloxy-$l^ 2-boranuidyl acetate
IUPAC Name sodium bis(acetyloxy)boranuidyl acetate
SMILES [Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O

Specifications

Melting Point ∼110°C (decomposition)
Quantity 25 g
UN Number UN3396
Beilstein 4047608
Sensitivity Moisture sensitive
Solubility Information Soluble in dimethyl sulfoxide,methanol,benzene,toluene,terahydrofuran,dioxane and methylene chloride.
Formula Weight 211.94
Percent Purity 95%
Chemical Name or Material Sodium triacetoxyborohydride
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium triacetoxyborohydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H228-Flammable solid.
  • H261-In contact with water releases flammable gases.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
Supplemental information
  • MIXTURE LIST-Contains : Sodium triacetoxyborohydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.