Learn More
Sodium triacetoxyborohydride, 95%
Description
Sodium triacetoxyborohydride is used as a reagent in reductive amination of ketones, aldehydes and lactamization of carbonyl compounds with amines. It also serves as a reducing agent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium triacetoxyborohydride is used as a reagent in reductive amination of ketones, aldehydes and lactamization of carbonyl compounds with amines. It also serves as a reducing agent in organic synthesis.
Solubility
Soluble in dimethyl sulfoxide, methanol, benzene, toluene, terahydrofuran, dioxane and methylene chloride.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, water, alcohols and acids.
Specifications
Specifications
| Melting Point | ∼110°C (decomposition) |
| Quantity | 5 g |
| UN Number | UN3396 |
| Beilstein | 4047608 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in dimethyl sulfoxide,methanol,benzene,toluene,terahydrofuran,dioxane and methylene chloride. |
| Formula Weight | 211.94 |
| Percent Purity | 95% |
| Chemical Name or Material | Sodium triacetoxyborohydride |
Safety and Handling
- Sodium triacetoxyborohydride
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Substance which, in contact with water, emits flammable gases Category 2
- H228-Flammable solid.
- H261-In contact with water releases flammable gases.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- MIXTURE LIST-Contain: Sodium triacetoxyborohydride
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.